Cerium(III) bromide, ultra dry, 99.99% (metals basis)
Cerium(III) bromide, ultra dry, 99.99% (metals basis)
Cerium(III) bromide, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Cerium(III) bromide, ultra dry, 99.99% (metals basis)

CAS: 14457-87-5 | Br3Ce | 379.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 047194.06
also known as 047194-06
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS14457-87-5
IUPAC Namecerium(3+) tribromide
Molecular FormulaBr3Ce
InChI KeyMOOUSOJAOQPDEH-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to gray
FormBead
Total Metal Impurities0.01% max.
Particle Size-10 Mesh
It is used as an additive for chemical reagent. Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only