Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)
Thermo Scientific Chemicals

Starch indicator solution 1%, w/v aqueous solution (for iodometric titrations)

For iodometric titrations | CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
20 L
Catalog number 047209.M4
also known as 047209-M4
Price (USD)
233.00
Each
Quantity:
20 L
Request bulk or custom format
Price (USD)
233.00
Each
Chemical Identifiers
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
FormLiquid
Concentration1.0% w/v ± 0.05% w/v
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only