Copper(I) bromide, 98%
Copper(I) bromide, 98%
Copper(I) bromide, 98%
Thermo Scientific Chemicals

Copper(I) bromide, 98%

Catalyst | CAS: 7787-70-4 | BrCu | 143.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Color:
Black-Gray
Green
Catalog number 047211.30
also known as 047211-30
Price (USD)
96.40
Each
Quantity:
250 g
Color:
Black-Gray
Request bulk or custom format
Price (USD)
96.40
Each
Chemical Identifiers
CAS7787-70-4
IUPAC Nameλ¹-copper(1+) bromide
Molecular FormulaBrCu
InChI KeyNKNDPYCGAZPOFS-UHFFFAOYSA-M
SMILES[Cu+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)>= 97.5%
Appearance (Color)light green
Formpowder
It finds many applications in organic synthesis, as a catalyst and a reagent. It is used as a catalyst in Sandmeyer reaction for preparing aryl bromides from arylamines through diazonium salts. It forms complexes with Lewis bases, such as dimethylsulfide, which is a precursor to organocopper reagents. Reactions involving Atom Transfer Radical Polymerization (ATRP) and Cu-catalyzed Cross-Dehydrogenative Couplings (CDC) are catalyzed by CuBr complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only