Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
Thermo Scientific Chemicals

Tris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate

CAS: 80049-61-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 047214.03
also known as 047214-03
Price (USD)
420.65
Special offer
Online exclusive
Ends: 28-Dec-2026
495.00
Save 74.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
420.65
Special offer
Online exclusive
Ends: 28-Dec-2026
495.00
Save 74.35
Each
Specifications
CAS80049-61-2
Chemical Name or MaterialTris(acetonitrile)cyclopentadienylruthenium(II) hexafluorophosphate
ColorYellow to Orange
Formula Weight434.28
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only