(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%
(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%
Thermo Scientific™

(S)-(4,4',5,5',6,6'-Hexamethoxybiphenyl-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphine), 97%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 047218.03
also known as 047218-03
Price (USD)
410.65
Online Exclusive
456.00
Save 45.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
410.65
Online Exclusive
456.00
Save 45.35 (10%)
Each
Add to cart
Specifications
Formula Weight814.92
Molecular FormulaC{5}{0}H{5}{6}O{6}P{2}
Synonym(S)-(4,4',5,5',6,6'-Hexamethoxy[1,1'-biphenyl]-2,2'-diyl)bis(bis[3,5-dimethylphenyl]phosphane);
Percent Purity97%
Physical FormSolid
View more