Copper, Oil based standard solution, Specpure™ Cu 5000μg/g
Copper, Oil based standard solution, Specpure™ Cu 5000μg/g
Copper, Oil based standard solution, Specpure™ Cu 5000μg/g
Thermo Scientific Chemicals

Copper, Oil based standard solution, Specpure™ Cu 5000μg/g

CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Catalog NumberQuantity
047225.18
also known as 047225-18
50 g
Catalog number 047225.18
also known as 047225-18
Price (USD)
82.90
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
82.90
Each
Specifications
CAS7697-37-2
ChEBICHEBI:48107
Chemical Name or MaterialCopper
IUPAC Namenitric acid
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only