Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
047229.KF
also known as 047229-KF
1 Pc.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047229.KF
also known as 047229-KF
Price (USD)
-
Quantity:
1 Pc.
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-69-9
ChEBICHEBI:33301
Chemical Name or MaterialBismuth rod
EINECS Number231-177-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only