Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH
Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH
Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH
Thermo Scientific Chemicals

Cerium(IV) oxide, 20% in H{2}O, nanoparticle dispersion, high pH

CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 047240.22
also known as 047240-22
Price (USD)
187.65
Special offer
Online exclusive
Ends: 28-Dec-2026
220.00
Save 32.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
187.65
Special offer
Online exclusive
Ends: 28-Dec-2026
220.00
Save 32.35
Each
Chemical Identifiers
CAS1306-38-3
IUPAC Namedioxocerium
Molecular FormulaCeO2
InChI KeyCETPSERCERDGAM-UHFFFAOYSA-N
SMILESO=[Ce]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration (weight/weight)20% min
The addition of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures. It is used as an additive in glass industry. Cerium dioxide is mainly used as such or in formulation as a high precision polishing agent for glass products, as catalyst or catalyst support. Polishing and decolorizing glass; opacifier in vitreous enamels and photochromic glasses; heat-resistant alloy coatings; as a cracking catalyst; as a catalyst for automobile emission control; in ceramic coatings; in phosphors; in cathodes; in capacitors; in semiconductors; in refractory oxides; gemstone polishing, as raw material for the production of glass and ceramics as well as additives in paints and varnishes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only