BrettPhos Palladacycle 3rd Generation, 98%
Thermo Scientific Chemicals

BrettPhos Palladacycle 3rd Generation, 98%

CAS: 1470372-59-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 047250.03
also known as 047250-03
Price (USD)
434.65
Special offer
Online exclusive
Ends: 30-Jun-2026
511.00
Save 76.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
434.65
Special offer
Online exclusive
Ends: 30-Jun-2026
511.00
Save 76.35 (15%)
Each
Chemical Identifiers
CAS1470372-59-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale brown
FormPowder
Elemental Analysis1.1 to 2.1 wt% Nitrogen
Elemental Analysis6.8 to 7.8 wt% Hydrogen
Elemental Analysis62.0 to 65.0 wt% Carbon
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only