Strontium iodide, anhydrous, 99% (metals basis)
Strontium iodide, anhydrous, 99% (metals basis)
Strontium iodide, anhydrous, 99% (metals basis)
Thermo Scientific Chemicals

Strontium iodide, anhydrous, 99% (metals basis)

CAS: 10476-86-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 047268.06
also known as 047268-06
Price (USD)
63.50
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
63.50
Each
Specifications
Assay Percent Notes(metals basis)
CAS10476-86-5
Chemical Name or MaterialStrontium iodide
ColorBrown to Yellow
Density4.550g/mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only