Barium iodide dihydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium iodide dihydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047314.A4
also known as 047314-A4
Price (USD)
615.65
Online Exclusive
684.00
Save 68.35 (10%)
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
615.65
Online Exclusive
684.00
Save 68.35 (10%)
Each
Chemical Identifiers
CAS7787-33-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormPowder or crystalline powder
Assay (Titration ex Iodide)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.