Cadmium borate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium borate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047321.30
also known as 047321-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Specifications
CAS51222-60-7
Molecular FormulaB{2}Cd{3}O{6}
SynonymBoric acid cadmium salt;
Percent Purity≥98%
Physical FormSolid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov