Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable AcroSeal™ bottles
Thermo Scientific™

Pyridine, anhydrous, 99.5+%, packaged under Argon in resealable AcroSeal™ bottles

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 047323.K2
also known as 047323-K2
Price (USD)
200.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
200.00
Each
Chemical Identifiers
CAS110-86-1
IUPAC Namepyridine
Molecular FormulaC5H5N
InChI KeyJUJWROOIHBZHMG-UHFFFAOYSA-N
SMILESC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay from Supplier's CofA≥99.5% (GC, corrected for water)
Optical Absorbance at330 nm: ≤1.00 AU
View more
WARNING: Cancer – www.P65Warnings.ca.gov