Potassium chromate, 99.9%
Potassium chromate, 99.9%
Potassium chromate, 99.9%
Thermo Scientific Chemicals

Potassium chromate, 99.9%

CAS: 7789-00-6 | CrK2O4 | 194.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047324.14
also known as 047324-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7789-00-6
IUPAC Namedipotassium dioxochromiumbis(olate)
Molecular FormulaCrK2O4
InChI KeyXMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES[K+].[K+].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Trace Metals (by ICP)≤ 0.1% (≤1,000ppm)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov