Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable AcroSeal™ bottles, stab. with amylene
Thermo Scientific™

Dichloromethane, anhydrous, 99.7+%, packaged under Argon in resealable AcroSeal™ bottles, stab. with amylene

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
4 x 1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 047352.K2
also known as 047352-K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)0.01% max
Residue on Evaporation3 ppm max
Assay (GC)99.7% min
WARNING: Cancer – www.P65Warnings.ca.gov