Barium perrhenate, Puratronic™, 99.99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium perrhenate, Puratronic™, 99.99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 047360.03
also known as 047360-03
Price (USD)
32.65
Online Exclusive
35.80
Save 3.15 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
32.65
Online Exclusive
35.80
Save 3.15 (9%)
Each
Specifications
CAS13768-52-0
Molecular FormulaBaO{8}Re{2}
Percent Purity99.99%
Physical FormSolid
Quantity1 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.