Potassium metaborate hydrate, 99+%
Potassium metaborate hydrate, 99+%
Thermo Scientific Chemicals

Potassium metaborate hydrate, 99+%

CAS: 16481-66-6 | KBO{2}^.xH{2}O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047365.A1
also known as 047365-A1
Price (USD)
77.65
Special offer
Online exclusive
Ends: 31-Dec-2025
90.80
Save 13.15 (14%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 31-Dec-2025
90.80
Save 13.15 (14%)
Each
Add to cart
Specifications
CAS16481-66-6
Melting Point950°C
Molecular FormulaKBO{2}^.xH{2}O
SynonymPotassium borate (meta)
Percent Purity≥99%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.