Bismuth(III) perchlorate oxide hydrate, 96%
Thermo Scientific Chemicals

Bismuth(III) perchlorate oxide hydrate, 96%

CAS: 66172-93-8 | BiOClO{4}^.xH{2}O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047372.30
also known as 047372-30
Price (USD)
103.65
Special offer
Online exclusive
Ends: 28-Dec-2026
122.00
Save 18.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 28-Dec-2026
122.00
Save 18.35
Each
Specifications
CAS66172-93-8
Molecular FormulaBiOClO{4}^.xH{2}O
SynonymBismuth oxyperchlorate hydrate;
Percent Purity96%
Physical FormSolid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.