Paraformaldehyde, 32% w/v aq. soln., methanol free
Paraformaldehyde, 32% w/v aq. soln., methanol free
Paraformaldehyde, 32% w/v aq. soln., methanol free
Thermo Scientific Chemicals

Paraformaldehyde, 32% w/v aq. soln., methanol free

Solution is micro-filtered and supplied in ampules. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 x 10 mL
10 x 10 mL
Catalog number 047377.9L
also known as 047377-9L
Price (USD)
57.65
Online Exclusive
64.00
Save 6.35 (10%)
Each
Quantity:
2 x 10 mL
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
64.00
Save 6.35 (10%)
Each
Chemical Identifiers
CAS50-00-0
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Concentration31.0-33% (Formaldehyde)
CommentAcidity (as formic): ≤0.01%
FormLiquid
Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.
RUO – Research Use Only