Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable AcroSeal™ bottles
Thermo Scientific™

Tetrahydrofuran, anhydrous, 99.8+%, stab. with 0.025% BHT, packaged under Argon in resealable AcroSeal™ bottles

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
4 x 1 L
Catalog number 047382.M1
also known as 047382-M1
Price (USD)
431.00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
431.00
Each
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.80% (corrected for water)
Acid ValueCH3COOH: ≤0.0005 meq/g (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
WARNING: Cancer – www.P65Warnings.ca.gov