Lithium chromate dihydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium chromate dihydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047383.30
also known as 047383-30
Price (USD)
73.65
Online exclusive
81.50
Save 7.85
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
73.65
Online exclusive
81.50
Save 7.85
Each
Chemical Identifiers
CAS7789-01-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay from Supplier's CofA≥96.0%
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov