Bismuth sub-nitrate monohydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth sub-nitrate monohydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Catalog number 047389.30
also known as 047389-30
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.70
Save 14.05
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.70
Save 14.05
Each
Specifications
CAS13595-83-0
Molecular FormulaBiONO{3}^.H{2}O
SynonymNitric acid dihydroxybismuthinyl ester
Percent Purity≥98%
Physical FormSolid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.