Cadmium acetate, anhydrous, 99+%
Thermo Scientific Chemicals

Cadmium acetate, anhydrous, 99+%

CAS: 89759-80-8 | C{4}H{6}CdO{4}
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number 047396.22
also known as 047396-22
Price (USD)
54.65
Special offer
Online exclusive
Ends: 28-Dec-2026
64.70
Save 10.05
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 28-Dec-2026
64.70
Save 10.05
Each
Specifications
CAS89759-80-8
Melting Point255°C
Molecular FormulaC{4}H{6}CdO{4}
SynonymAcetic acid cadmium salt;
Percent Purity≥99%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov