Molybdenum(IV) oxide, 99% (metals basis)
Molybdenum(IV) oxide, 99% (metals basis)
Molybdenum(IV) oxide, 99% (metals basis)
Thermo Scientific Chemicals

Molybdenum(IV) oxide, 99% (metals basis)

CAS: 18868-43-4 | MoO2 | 127.948 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 048117.22
also known as 048117-22
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS18868-43-4
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only