Niobium(V) chloride, 99% (metals basis)
Niobium(V) chloride, 99% (metals basis)
Niobium(V) chloride, 99% (metals basis)
Thermo Scientific Chemicals

Niobium(V) chloride, 99% (metals basis)

For Use in Preparation of Pure Niobium, and As An Intermediate | CAS: 10026-12-7 | Cl5Nb | 270.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 051108.22
also known as 051108-22
Price (USD)
217.65
Special offer
Online exclusive
Ends: 30-Jun-2026
256.00
Save 38.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
217.65
Special offer
Online exclusive
Ends: 30-Jun-2026
256.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS10026-12-7
IUPAC Nameniobium(5+) pentachloride
Molecular FormulaCl5Nb
InChI KeyYHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
In preparation of pure niobium, and as an intermediate.Niobium(V) chloride is the precursor in the preparation of pure niobium, and ferroniobium. It is used as Lewis acid catalyst in the organic synthesis in activating alkenes in the carbonyl-ene reaction and the Diels-Alder reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only