Dionex™ AMC-1 Anion Micro Concentrator Column for IC
Thermo Scientific™

Dionex™ AMC-1 Anion Micro Concentrator Column for IC

Green features
Concentrate inorganic and low molecular weight organic anions from ultrapure water with this column featuring novel solvent-compatible resin technology.
Have Questions?
Catalog NumberLength (Metric)
05176015 mm
Catalog number 051760
Price (MXN)
-
Length (Metric):
15 mm

Concentrate inorganic and low molecular weight organic anions from ultrapure water with the Thermo Scientific™ Dionex™ AMC-1 Anion Microconcentrator Column. This column has a novel solvent-compatible resin technology that ensures a low sulfate background during the concentration step. The low void volume (∼15μL) ensures accurate determination of early eluting anions such as fluoride, glycolate, acetate, and formate. It also reduces the system dip and improves anion determinations for trace anions in the power generation and semiconductor industries.

Reduce Detection Limits in Water Analysis by 2–5 Orders of Magnitude

Dionex IonPac and Thermo Scientific™ Dionex™ IonSwift™ anion concentrator columns are designed primarily for high-purity water analysis. The concentrator retains ions from a measured volume of aqueous sample matrix, concentrating the analyte species and lowering detection limits by 2–5 orders of magnitude.

Engineered for Superior Chromatographic Performance
  • Backpressure operation (<400psi at 2mL/min)
  • Recommended for carbonate/bicarbonate and borate eluents
  • 3.0μeq/column capacity
  • Low void volume (∼15μL)
  • Single format (2 x 15mm)
  • Ideal for 2mm concentration methods
  • Compatible with a single-piston sample delivery pump (Thermo Scientific Dionex AXP Auxiliary Pump)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity3 μeq
Column ChemistryCarbonate/Bicarbonate/Borate
Crosslinking55% DVB
DescriptionAMC-1; 2 x 15 mm
Diameter (Metric) Latex10 μm
Flow Rate0.1 to 2.0 mL/min.
For Use With (Application)Concentration of Inorganic and Low-Molecular-Weight Organic Anions from Ultrapure Water, Anion Microconcentrator
For Use With (Equipment)2 mm or 4 mm I.D. Anion-Exchange Systems
Green FeaturesFSC
HydrophobicityMedium
Max. Pressure3000 psi (207 bar)
Particle Size10 μm
QuantityEach 1
Solvent CompatibilityCarbonate/Bicarbonate or Borate Eluents
TypeAnion Microconcentrator Column
Void Volume (Metric)15 μL
Diameter (Metric)2 mm
Length (Metric)15 mm
Unit SizeEach 1