Nickel(II) tetrafluoroborate hexahydrate
Nickel(II) tetrafluoroborate hexahydrate
Nickel(II) tetrafluoroborate hexahydrate
Thermo Scientific Chemicals

Nickel(II) tetrafluoroborate hexahydrate

CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 053108.22
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS15684-36-3
IUPAC Namenickel(2+) bis(tetrafluoroboranuide) hexahydrate
Molecular FormulaB2F8H12NiO6
InChI KeyBFFWGFMAXHCLDV-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ni > 16.73%
It is mainly used as an electroplating intermediate. It is also used in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only