Nickel(II) sulfate hexahydrate, 99.97% min (metals basis)
Nickel(II) sulfate hexahydrate, 99.97% min (metals basis)
Nickel(II) sulfate hexahydrate, 99.97% min (metals basis)
Nickel(II) sulfate hexahydrate, 99.97% min (metals basis)
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 99.97% min (metals basis)

CAS: 10101-97-0 | H12NiO10S | 262.839 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 053130.30
also known as 053130-30
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
400.00
Save 59.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
400.00
Save 59.35 (15%)
Each
Chemical Identifiers
CAS10101-97-0
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
Nickel(II) sulfate hexahydrate is mainly used in Ni plating, as mordant in dyeing and in printing textiles, as a blackening agent for zinc and brass and in the fabrication of organic nickel salts. It is used as an effective catalyst for gas-phase dehydration of glycerol to acrolein. Nickel sulfate hexahydrate crystal is an efficient ultraviolet filter material. Columns used in biochemistry and molecular biology can be regenerated with nickel sulfate. It is used to prepare nickel carbonate, a precursor to pigments and nickel-based catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only