Nickel(II) chloride hexahydrate, 99.95% (metals basis)
Nickel(II) chloride hexahydrate, 99.95% (metals basis)
Nickel(II) chloride hexahydrate, 99.95% (metals basis)
Nickel(II) chloride hexahydrate, 99.95% (metals basis)
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 99.95% (metals basis)

CAS: 7791-20-0 | Cl2Ni·6H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 053131.A1
also known as 053131-A1
Price (USD)
456.65
Online Exclusive
507.00
Save 50.35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
456.65
Online Exclusive
507.00
Save 50.35 (10%)
Each
Chemical Identifiers
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
FormCrystalline powder or crystals
Assay from Supplier's CofA≥99.95% metals basis
Total Metal Impurities≤0.05% (500 ppm)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only