Dionex™ PC10 Postcolumn Pneumatic Delivery Packages
Actual product may vary
Thermo Scientific™

Dionex™ PC10 Postcolumn Pneumatic Delivery Packages

Ensure smooth, optimal reagent flow for applications with high background absorbances, such as trace-level transition metal analysis using post-column reaction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
0535912 mm
0506014 mm
Catalog number 053591
Price (USD)
4,470.00
Each 1
Diameter (Metric):
2 mm
Price (USD)
4,470.00
Each 1
Ensure consistent, pulse-free reagent delivery for applications with high background absorbances, such as trace-level transition metal analysis using post-column reaction with 4-(2-pyridylazo)resorcinol (PAR). The Thermo Scientific™ Dionex™ PC10 Postcolumn Pneumatic Delivery Package provides reagent with less pressure pulsation by applying constant pressure, eliminating the need for a pump.

The PC10 Postcolumn Pneumatic Delivery System ensures a constant, pulseless flow of reagent. It assures optimal flow by applying constant pressure, eliminating the need for a pump and its associated moving parts, labor and expense.

  • Offered as 2 or 4mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (Metric)2 mm
QuantityEach 1
TypePostcolumn Pneumatic Delivery Pk.
For Use With (Equipment)PC10 Postcolumn Delivery System
Unit SizeEach 1