Sodium peroxydisulfate, 98%
Sodium peroxydisulfate, 98%
Sodium peroxydisulfate, 98%
Sodium peroxydisulfate, 98%
Thermo Scientific Chemicals

Sodium peroxydisulfate, 98%

CAS: 7775-27-1 | Na2O8S2 | 238.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 054100.A7
also known as 054100-A7
Price (USD)
148.65
Special offer
Online exclusive
Ends: 30-Jun-2026
175.00
Save 26.35 (15%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
148.65
Special offer
Online exclusive
Ends: 30-Jun-2026
175.00
Save 26.35 (15%)
Each
Chemical Identifiers
CAS7775-27-1
IUPAC Namedisodium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaNa2O8S2
InChI KeyCHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (special)≥ 97.5%
FormCrystalline
Appearance (Color)White
Sodium peroxydisulfate is used as a radical initiator for emulsion polymerization reactions like acrylonitrile butadiene styrene, detergent component, soil conditioner and soil remediation. It is also used for curing of formaldehyde adhesives. It acts as a bleaching agent and in the production of dyestuffs. It finds application in zinc and printed circuit boards and considered to be a replacement for ammonium persulfate in etching mixtures. Further, it is used in the preparation of diapocynin from apocynin. It plays a role in the conversion of phenols to para-diphenols in alkaline solution and of arylamines to aminophenols. It is actively involved in the Elbs persulfate oxidation and the Boyland-Sims oxidation reactions as an oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only