Product Image
Thermo Scientific™
Dionex™ PEEK Pump Kits
Thermo Scientific™ PEEK Pump Kits are for use with the GP50/IP25 Standard Bore, GP50/IP25 Microbore and GS50/IS25.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
055639GP50/IP25 Standard Bore
055641GP50/IP25 Microbore
060685GS50/IS25
Catalog number 055639
Price (CLP)
-
Description:
GP50/IP25 Standard Bore
Thermo Scientific™ PEEK Pump Kits are for use with the GP50/IP25 Standard Bore, GP50/IP25 Microbore and GS50/IS25.
Each maintenance kit includes the required spare parts to perform the recommended annual maintenance on the specified module.
Specifications
DescriptionGP50/IP25 Standard Bore
For Use With (Application)Ion Chromatography
MaterialPEEK
For Use With (Equipment)GP50/IP25 Standard Bore
Product LineDionex
TypePump Kit
Unit SizeEach