12-Molybdophosphoric acid hydrate, ACS
12-Molybdophosphoric acid hydrate, ACS
12-Molybdophosphoric acid hydrate, ACS
Thermo Scientific Chemicals

12-Molybdophosphoric acid hydrate, ACS

CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 056166.14
also known as 056166-14
Price (USD)
75.65
Online Exclusive
83.70
Save 8.05 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
83.70
Save 8.05 (10%)
Each
Chemical Identifiers
CAS51429-74-4
IUPAC Name12-Molybdophosphoric acid
Molecular FormulaH3Mo12O40P
Molecular Weight (g/mol)1825.40
Synonymphosphoric acid dodecamolybdenum hexatriacontaoxidandiide
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.02%
Impurity contentCl 0.02%
Impurity contentFe 0.005%
Impurity contentNH₄ 0.01%
Impurity contentSO₄ 0.025%
View more
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only