Product Image
Thermo Scientific™

Dionex™ OnGuard™ II RP Cartridges

Remove hydrophobic substances, aromatic carboxylic acids, hydrocarbons, and surfactants from sample to improve IC performance. Pretreat 12 samples in parallel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
0570841
082760OnGuard II RP Cartridge, 1-cc12
0570831
Catalog number 057084
Price (USD)
680.00
Each 1
Quantity:
1
Price (USD)
680.00
Each 1

Remove hydrophobic substances such as aromatic dyes, some aromatic carboxylic acids, hydrocarbons, and surfactants from sample matrices to improve IC performance and facilitate low-level ion analysis. Thermo Scientific™ Dionex™ OnGuard™ II RP Cartridges are an off-line sample preparation technique that uses a macroporous divinylbenzene resin with very high selectivity for hydrophobic substances to eliminate these matrix interferences. Use these cartridges singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific™ Dionex™ OnGuard™ Sample Prep Station.

Solve Tough Application Problems

  • Boost sensitivity, lower baselines, and get better separations by removing phenolics, metals, cations, anions, hydrophobic substances, and more
  • Achieve reproducible trace-level determinations in concentrated matrices
  • Increase the lifetime of your analytical columns
  • Use for a wide range of matrices thanks to wide pH stability
  • Supports applications with choice of 1cc and 2.5cc formats, as well as syringe barrel format for robotic processing (custom order)

Easy-to-Use, Leak-Free

  • Leak-free, no-channeling cartridge
  • Quick, secure Luer inlet connections

Performance Guaranteed

  • Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness

Pretreat 12 Samples Simultaneously

  • Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
DescriptionRP; 2.5 cc
For Use With (Application)Removal of Hydrophobic Substances
MaterialMacroporous Divinylbenzene Resin
Quantity1
Volume (Metric)2.5 mL
For Use With (Equipment)Dionex OnGuard Sample Prep Station
Product LineOnGuard
pH0 to 14
Unit SizeEach 1