Product Image
Thermo Scientific™

Dionex™ OnGuard™ II Ag Cartridges

Remove chloride, bromide, and iodide from sample matrices off-line to improve IC performance and facilitate trace analyses. Pretreat up to 12 samples in parallel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
057089OnGuard II Ag Cartridge, 1-cc48
082762OnGuard II Ag Cartridge, 1-cc12
057090OnGuard II Ag Cartridge, 2.5-cc48
Catalog number 057089
Price (USD)
407.00
Each 1
Type:
OnGuard II Ag Cartridge, 1-cc
Quantity:
48
Price (USD)
407.00
Each 1
Remove chloride, bromide, and iodide from sample matrices to improve performance of ion chromatography (IC) and facilitate trace analyses. Performed prior to IC analysis, the Thermo Scientific™ Dionex™ OnGuard™ II Ag Cartridge is an off-line sample preparation technique that eliminates these matrix interferences using a silver-form, high-capacity, suffocated, cation-exchange resin. Pretreat a single sample or many samples in series. Use them singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific™ Dionex™ OnGuard™ Sample Prep Station.
Dionex OnGuard II Ag Cartridge has a similar resin to and is often used in concert with the Thermo Scientific™ Dionex™ OnGuard™ II H cartridge, which removes dissolved silver and other cations. Use the Dionex OnGuard II Ag Cartridge before using the Dionex™ OnGuard II H cartridge. Also consider the Thermo Scientific™ Dionex™ OnGuard™ II Ag/H cartridge.

Solve Tough Application Problems

  • Boost sensitivity, lower baselines, and get better separations
  • Achieve reproducible trace-level determinations in concentrated matrices
  • Increase the lifetime of your analytical columns
  • Use for a wide range of matrices thanks to wide pH stability
  • Supports applications with choice of 1cc and 2.5cc formats, as well as syringe barrel format for robotic processing (custom order)

Easy-to-Use, Leak-Free

  • Leak-free, no-channeling cartridge
  • Quick, secure Luer inlet connections
Performance Guaranteed
  • Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness
Pretreat 12 Samples Simultaneously
  • Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
DescriptionCation-exchange silver form
For Use With (Application)Removal of Chloride, Bromide, and Iodide from Sample Matrices
MaterialSilver-Form, High-Capacity, Suffocated, Cation-Exchange Resin
Quantity48
Sample Capacity2.0 to 2.2 (1 cc); 5.0 to 5.5 (2.5 cc)
Volume (Metric)1 mL
For Use With (Equipment)Dionex OnGuard Sample Prep Station
Product LineOnGuard
TypeOnGuard II Ag Cartridge, 1-cc
pH0 to 14
Unit SizeEach 1