Product Image
Thermo Scientific™

Dionex™ OnGuard™ II Ag Cartridges

Remove chloride, bromide, and iodide from sample matrices off-line to improve IC performance and facilitate trace analyses. Pretreat up to 12 samples in parallel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
05709048
08276212
057089OnGuard II Ag Cartridge, 1-cc48
Catalog number 057090
Price (USD)
886.00
Each
Quantity:
48
Price (USD)
886.00
Each
Remove chloride, bromide, and iodide from sample matrices to improve performance of ion chromatography (IC) and facilitate trace analyses. Performed prior to IC analysis, the Thermo Scientific™ Dionex™ OnGuard™ II Ag Cartridge is an off-line sample preparation technique that eliminates these matrix interferences using a silver-form, high-capacity, suffocated, cation-exchange resin. Pretreat a single sample or many samples in series. Use them singly or in series, or pretreat up to 12 samples in parallel using the optional Thermo Scientific™ Dionex™ OnGuard™ Sample Prep Station.
Dionex OnGuard II Ag Cartridge has a similar resin to and is often used in concert with the Thermo Scientific™ Dionex™ OnGuard™ II H cartridge, which removes dissolved silver and other cations. Use the Dionex OnGuard II Ag Cartridge before using the Dionex™ OnGuard II H cartridge. Also consider the Thermo Scientific™ Dionex™ OnGuard™ II Ag/H cartridge.

Solve Tough Application Problems

  • Boost sensitivity, lower baselines, and get better separations
  • Achieve reproducible trace-level determinations in concentrated matrices
  • Increase the lifetime of your analytical columns
  • Use for a wide range of matrices thanks to wide pH stability
  • Supports applications with choice of 1cc and 2.5cc formats, as well as syringe barrel format for robotic processing (custom order)

Easy-to-Use, Leak-Free

  • Leak-free, no-channeling cartridge
  • Quick, secure Luer inlet connections
Performance Guaranteed
  • Ships with a Certificate of Analysis that verifies capacity, performance and cleanliness
Pretreat 12 Samples Simultaneously
  • Use the optional Dionex OnGuard Sample Prep Station and OnGuard Sample Pretreatment Cartridges
Specifications
DescriptionCation-exchange silver form
For Use With (Application)Removal of Chloride, Bromide, and Iodide from Sample Matrices
MaterialSilver-Form, High-Capacity, Suffocated, Cation-Exchange Resin
Quantity48
Sample Capacity2.0 to 2.2 (1 cc); 5.0 to 5.5 (2.5 cc)
Volume (Metric)2.5 mL
For Use With (Equipment)Dionex OnGuard Sample Prep Station
Product LineOnGuard
pH0 to 14
Unit SizeEach