Product Image
Thermo Scientific™

Dionex™ CarboPac™ PA1 IC Columns

Green features
For isocratic determinations of linear polysaccharides and mono- and disaccharides in a variety of matrices, use Thermo Scientific™ Dionex™ CarboPac™ PA1 Analytical & Guard Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColumn TypeLength (Metric)Diameter (Metric)
057179Guard Column50 mm2 mm
057178Analytical Column250 mm2 mm
043096Guard Column50 mm4 mm
039686Semi-Preparative Column250 mm9 mm
063501Semi-Preparative Guard Column50 mm9 mm
035391Analytical Column250 mm4 mm
Catalog number 057179
Price (USD)
631.00
Each 1
Column Type:
Guard Column
Length (Metric):
50 mm
Diameter (Metric):
2 mm
Price (USD)
631.00
Each 1
For isocratic determinations of monosaccharides, disaccharides, and linear polysaccharides in a variety of matrices, use the rugged Thermo Scientific™ Dionex™ CarboPac™ PA1 Analytical & Guard Columns. This specialized anion-exchange column, combined with pulsed amperometric detection (PAD), delivers high-resolution separations. The unique Thermo Scientific™ Dionex™ MicroBead™ pellicular structure of the Dionex CarboPac PA1 resin gives it stability from pH 0–14 at all concentrations of buffer salts. The pellicular resin structure provides excellent mass transfer resulting in fast gradient re-equilibration.
Durable Column for Carbohydrate Separations
  • Approved for use in a variety of official methods for the analysis of foods
  • Well suited for the analysis of foods for nutritional labeling
  • Ideal for the separation of neutral and acidic monosaccharides
  • Lower sensitivity than the Dionex CarboPac PA10 or PA20 Columns
  • Available in microbore, standard bore, and semipreparative formats

About the Dionex CarboPac Family of Columns

The Dionex CarboPac family offers a selection of high-performance liquid chromatography (HPLC) columns, each optimized for a different class of compounds. Combined with PAD, which provides a means to detect carbohydrates without derivatization, these columns support simple, reliable techniques to separate sugar alcohols, mono- and disaccharides, and poly- and oligosaccharides.

Dionex CarboPac columns provide high-resolution separations of:

  • Glycoprotein oligosaccharides and complex carbohydrates from dietary fiber, including fructans, maltodextrins, and amylopectins
  • Monosaccharides, such as those obtained from glycoprotein hydrolysates or plant hydrolysates
  • Mono- and disaccharides
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAOAC Methods 995.13, 996.04, 997.08, 2000.11, 2000.17 and 2001.02
Column Capacity5 μeq
For Use With (Application)Separation of Neutral and Acidic Monosaccharides, Carbohydrate Analysis
For Use With (Equipment)Separation of Neutral, Acidic Monosaccharides
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
Particle Size10 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14, Up to 2% of Common HPLC Solvents
Stationary PhaseCarboPac PA1
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Unit SizeEach 1