Dionex™ AAA-Direct Reagents
Actual product may vary
Dionex™ AAA-Direct Reagents
Dionex™ AAA-Direct Reagents
Thermo Scientific™
Dionex™ AAA-Direct Reagents
Ensure excellent performance from your Thermo Scientific™ Dionex™ AAA-Direct system with these high-grade, pre-qualified reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
059540Histidine Standard
059326Sodium Acetate Salt
Catalog number 059540
Price (USD)
96.10
Each 1
Description:
Histidine Standard
Price (USD)
96.10
Each 1
High quality sodium acetate eluent and reliable histidine standards are crucial to reliable amino acid analysis. Ensure the best possible results by using Thermo Scientific™ Dionex™ AAA-Direct Reagents with your Dionex™ AAA-Direct system. Reagents include high-grade, contaminant-free sodium acetate and reliable histidine standard are pre-qualified and guaranteed not to contaminate your system. The sodium acetate is preweighed for convenient mixing —all you need to do is add water to produce the correct concentration of eluent.
  • High-purity sodium acetate for reliable amino acid results
  • Specifically tested for amino acid analysis
  • Ships with a certificate of analysis
  • Ships as a 2-pack of sodium acetate sufficient
  • Each container makes 1L of eluent for 1 month of continuous operation
  • Includes reliable histidine standard
  • Time-saving, pre-mixed standard
About the Dionex AAA-Direct System
The Dionex AAA-Direct system is a highly sensitive and direct-detection type of amino acid analyzer that requires no sample derivatization. The Dionex AAA-Direct ion chromatography system uses integrated pulsed amperometry (IPAD)—a type of electrochemical detection—to directly detect amino acids. The Dionex AAA-Direct also provides complete separation of all common amino acids using anion-exchange chromatography (Thermo Scientific™ Dionex™ AminoPac™ PA10 column).
Note: AAA-Direct users are recommended to purchase only the prequalified sodium acetate to guarantee trouble-free use of their system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionHistidine Standard
For Use With (Application)AAA-Direct System Installation
For Use With (Equipment)AAA-Direct System
HazardsIrritant
IncludesCertificate of Analysis
QuantityEach 1
Standard ComponentsL-Histidine HCl
Standard Concentration8 μM
StorageRoom Temperature
TypeAAA-Direct Reagents
Unit SizeEach 1