Dionex™ IonPac™ AS18 IC Columns
Imagem ilustrativa. O produto pode variar
Dionex™ IonPac™ AS18 IC Columns
Dionex™ IonPac™ AS18 IC Columns
Dionex™ IonPac™ AS18 IC Columns
Dionex™ IonPac™ AS18 IC Columns
Dionex™ IonPac™ AS18 IC Columns
Dionex™ IonPac™ AS18 IC Columns
Dionex™ IonPac™ AS18 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS18 IC Columns

Green features
Analyze inorganic anions and low-molecular-weight organic acids in complex matrices in less than 10 minutes using these 250 mm high capacity, hydroxide-selective anion-exchange columns.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoLength (Metric)Diameter (Metric)Column Type
06055150 mm4 mmGuard Column
060549250 mm4 mmAnalytical Column
060553250 mm2 mmAnalytical Column
06055550 mm2 mmGuard Column
075772250 mm0.4 mmAnalytical Column
07577350 mm0.4 mmGuard Column
Número do catálogo 060551
Preço (BRL)
-
Length (Metric):
50 mm
Diameter (Metric):
4 mm
Column Type:
Guard Column
Use these longer 250 mm Thermo Scientific™ Dionex™ IonPac™ AS18 columns higher resolution separations of more complex sample matrices. Dionex IonPac AS18 columns are high capacity hydroxide-selective anion-exchange columns. They are the column of choice for monitoring inorganic anions in drinking water and wastewater samples for compliance with U.S. EPA Methods 300.0 (A) and 300.1. This column is also recommended when separating the common inorganic anions and low-molecular weight organic acids in diverse sample matrices including drinking water, wastewater, leachates, foods, beverages, pharmaceutical preparations, and scrubber solutions.

High Resolution of Complex Sample Matrices

  • Longer 250 mm Dionex IonPac AS18 hydroxide-selective columns provide higher resolution separations of more complex sample matrices
  • Available in standard bore, microbore, and capillary formats Analysis of Common Inorganic Anions and Low Molecular Weight Organic

Acids in Diverse Matrices

  • Compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A)
  • Determination of low levels of inorganic anions in complex matrices using large-loop injections
  • Superior retention and quantification of fluoride, acetate, and formate
  • Isocratic separation of common inorganic anions in 9 min in diverse matrices using a 250 mm column
  • Higher-capacity applications use 250 mm columns allowing injection of more concentrated samples without overloading or peak broadening
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, or allow effective column cleanup

Recommended for Inorganic Anions in Diverse Sample Matrices

  • Source water and drinking water, municipal and industrial wastewater
  • Industrial cooling water, hazardous waste extracts and dump site leachates, acid rain
  • Foods and beverages, anionic counterions in pharmaceutical preparations and synthetic peptides
  • Polymers such as polyols and polysulfonates, scrubber solutions

Multiple 250 mm Formats for Application Flexibility

Get higher resolution separations of more complex sample matrices using these longer 250 mm analytical columns. Choose from standard bore, microbore, and capillary formats and use the corresponding guard column to protect and increase the life of your analytical column. Use the:

  • 4 x 250 mm standard bore column with the 4 x 50 mm guard column
  • 2 x 250 mm microbore columns with the 2 x 50 mm guard column
  • 0.4 x 250 mm capillary columns with the 0.4 x 50 mm guard column

Capillary Columns Reduce Operating Costs from Lower Eluent Requirements

Capillary columns provide significantly reduced operating and waste disposal costs through eluent conservation. Packed with the same material, the Dionex IonPac AS18 capillary columns provide the same performance as the standard bore and microbore counterparts. Operating costs are reduced because the capillary system operates at only 1% of the eluent flow rate of the standard bore system. Capillary columns are used with the Thermo Scientific™ Dionex™ ICS-4000 and ICS-5000+ Capillary systems.

Get Faster, High-Resolution separations from Dionex IonPac AS18-Fast and Dionex IonPac AS18-4μm Columns

  • For faster runs on relatively clean matrices, refer to the shorter 150 mm Dionex™ IonPac™ AS18-Fast Hydroxide-Selective Anion-Exchange Columns
  • For fastest, highest resolution separations, refer to the small particle Dionex™ IonPac™ AS18-4μm Hydroxide-Selective Anion-Exchange Columns. Use of 4μm columns requires a high-pressure IC system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Especificações
Certifications/ComplianceU.S. EPA Method 300.0 (A)
For Use With (Application)Anion Exchange
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Column TypeGuard Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)50 mm
Unit SizeEach 1