Product Image
Thermo Scientific™
Dionex™ Kit of Standards for OQ/PQ Measurements
The Thermo Scientific™ Dionex Kit of Standards is used for performance validation in ESI and APCI modes.
Catalog NumberStandard Concentration
0614961 mg/L
Catalog number 061496
Price (GBP)
-
Standard Concentration:
1 mg/L
The Thermo Scientific™ Dionex Kit of Standards is used for performance validation in ESI and APCI modes.
Includes:
Two standards, one for each mode.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)OQ/PQ Measurement
IncludesKit of 2 standards for performance validation in ESI and APCI modes
OperationUsed for performance validation in ESI and APCI modes.
Standard ComponentsPerchlorate
Standard Concentration1 mg/L
TypeStandard
Unit SizeEach