Dionex™ Rheodyne™ Valve Assemblies and Replacement Valve Pods
Actual product may vary
Thermo Scientific™
Dionex™ Rheodyne™ Valve Assemblies and Replacement Valve Pods
Simplify valve maintenance or facilitate switching between advanced IC applications with Dionex™ Rheodyne™ injection valve assemblies and replacement pods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
0619476-port Valve PodDionex Aquion IC systems
06196210-port Valve AssemblyDionex ICS-5000 systems
0746994-port Valve PodDionex ICS-4000 and ICS-6000 capillary IC systems
06194810-port Valve PodDionex Aquion IC systems
Catalog number 061947
Price (USD)
3,140.00
Each
Description:
6-port Valve Pod
For Use With (Equipment):
Dionex Aquion IC systems
Price (USD)
3,140.00
Each
Simplify valve maintenance or facilitate switching between advanced ion chromatography applications with Dionex™ Rheodyne™ injection valve assemblies and replacement valve pods. Valve assemblies include the valve actuator while valve pods are designed to use the existing actuator in the IC system.
Valve pods include the valve rotor and stator, and can easily be removed and installed into existing valve actuators. Valve pods for capillary applications include an internal sample loop. Valve assemblies include the valve pod and actuator.
Specifications
Description6-port Valve Pod
For Use With (Application)Ion Chromatography
For Use With (Equipment)Dionex Aquion IC systems
Product LineRheodyne
Unit SizeEach