Sodium perrhenate, 99.95% (metals basis)
Sodium perrhenate, 99.95% (metals basis)
Sodium perrhenate, 99.95% (metals basis)
Thermo Scientific™

Sodium perrhenate, 99.95% (metals basis)

CAS: 13472-33-8 | O3Re | 234.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 062107.03
also known as 062107-03
Price (USD)
74.50
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
74.50
Each
Chemical Identifiers
CAS13472-33-8
IUPAC Namesodium rheniumoylolate
Molecular FormulaNaO4Re
InChI KeyKMISVFTVBOPTAI-UHFFFAOYSA-N
SMILES[Na+].[O-][Re](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only