Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Actual product may vary
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns
Thermo Scientific™

Dionex™ IonPac™ Fast Anion IIIA Analytical & Guard Columns

Green features
Analyze phosphoric and citric acids in cola soft drink samples in less than 7 minutes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Column Type
062964250 mmAnalytical Column
06296650 mmGuard Column
Catalog number 062964
Price (CLP)
-
Length (Metric):
250 mm
Column Type:
Analytical Column
Analyze phosphoric and citric acids in cola soft drink samples in less than 7 minutes with the Thermo Scientific™ Dionex™ IonPac™ Fast Anion IIIA column. This rugged hydroxide-selective anion-exchange column delivers rapid, reliable results using an isocratic potassium hydroxide eluent delivered by an eluent generator in combination with suppressed conductivity detection.
Fast, Reliable Results With Simple Operation
  • Use with the eluent generator for simplified Reagent-Free IC (RFIC™) operation
  • Ensure low background and enhanced sensitivity by using the Thermo Scientific™ Dionex™ Anion Self-Regenerating Suppressor (ASRS™ 300)
  • Optimized for a 30˚C operating temperature to ensure reproducible retention times in all environmental conditions
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, or allow effective column cleanup
  • Economical operation provided by 3 mm i.d. configuration
  • Pretreatment
Cola samples may require sample pretreatment with sonication to remove carbonation. Sample dilution of cola syrups is recommended prior to analysis to ensure optimum column life.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity55 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
DescriptionFast Anion IIIA; 3 x 250 mm
Flow Rate1.0 to 2.0 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)Eluent Generator
Green FeaturesFSC
HydrophobicityUltralow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure3000 psi (207 bar)
Packing MaterialPolymeric
Particle Size7.5 μm
Pore Size2000 &‌Aring;
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Stationary PhaseSuper Macroporous Resin
Column TypeAnalytical Column
Diameter (Metric)3 mm
FormatMicrobore
Length (Metric)250 mm
Unit SizeEach 1