Dionex™ IonPac™ AS20 IC Columns
Actual product may vary
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Dionex™ IonPac™ AS20 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS20 IC Columns

Green features
Simplify trace analysis of perchlorate in drinking water, surface water, and groundwater matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
063148250 mm4 mmAnalytical Column
063065250 mm2 mmAnalytical Column
06306650 mm2 mmGuard Column
06315450 mm4 mmGuard Column
075399250 mm0.4 mmAnalytical Column
07540050 mm0.4 mmGuard Column
Catalog number 063148
Price (USD)
1,930.00
Each 1
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each 1
Simplify trace analysis of perchlorate in drinking water, surface water, and groundwater matrices using the Thermo Scientific™ Dionex™ IonPac™ AS20 Capillary, Analytical & Guard Columns. The capacity and selectivity of this column ensures that perchlorate can be quantified at low μg/L concentrations using suppressed conductivity detection, even in the presence of very high concentrations of chloride, carbonate, and sulfate. This column is specified in U.S. EPA Method 314.1 for the determination of trace perchlorate in drinking water (Confirmatory Method) after sample preconcentration.
Reduce Operating Costs With our New Capillary Column

The Dionex IonPac AS20 Capillary column is packed with the same material as the equivalent standard bore version (providing the same performance as a 4mm column) but requires only 1% of the eluent flow rate. The capillary format provides the advantage of significantly reduced eluent consumption, which helps reduce operating costs.

Superior Chromatographic Performance
  • Recommended hydroxide-selective, anion-exchange column for trace perchlorate in environmental water samples
  • High Capacity: 310μeq per column. (4 x 250 mm)
  • Increased mass sensitivity, reduced eluent consumption, and reduced operating costs
  • Operates at ambient or elevated temperature; column selectivity is optimized for 35°C operating temperature
  • Compatible with organic solvents
For Best Results
  • Use this column with a Reagent-Free™ IC (RFIC™) system and an eluent generator for optimum detection limits.
  • Use the Thermo Scientific™ Dionex™ Anion Electrolytically Regenerated Suppressor (AERS™ 500) or the Thermo Scientific™ Dionex™ Anion Capillary Electrolytic Suppressor (ACES™ 300) with the Dionex IonPac AS20 column for eluent suppression and enhanced analyte sensitivity.
  • Use the Thermo Scientific™ Cryptand C1 Concentrator column for sample preconcentration for analysis of trace perchlorate with the analytical column.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity310 μeq
Certifications/ComplianceU.S. EPA Method 314.1 (Confirmatory Method)
Column Capacity310 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
Flow Rate1.0 to 2.0 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)RFIC Systems
Green FeaturesFSC
HydrophobicityUltralow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure4000 psi (275 bar)
Packing MaterialPolymeric
Particle Size7.5 μm
Pore Size2000 &‌Aring;
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Stationary PhaseIonPac AS20
Column TypeAnalytical Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)250 mm
Unit SizeEach 1