ProPac™ HIC-10 HPLC Columns
Thermo Scientific™

ProPac™ HIC-10 HPLC Columns

Green features

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
06366575 mm7.8 mm
063653100 mm2.1 mm
063655100 mm4.6 mm
074197250 mm4.6 mm
SP6800150 mm10 mm
SP6869250 mm22 mm
Catalog number 063665
Price (USD)
1,905.75
Each
Length (Metric):
75 mm
Diameter (Metric):
7.8 mm
Price (USD)
1,905.75
Each

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns. Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.

High-efficiency Columns for the Separation of Large Biomolecules
  • High protein loading capacity for protein purification applications
  • Wide range of analytical and preparative applications, including the separation of bovine serum proteins, snake venom proteins, and monoclonal antibodies (Mabs)
  • Based on 5μm ultra-high purity spherical silica gel particles with 300Å pores

Hydrophobic interaction chromatography separates biomolecules in a decreasing salt gradient, based on differences in surface hydrophobicity. This method preserves the biological activity of proteins. The HIC separation mechanism is complementary to those of ion-exchange and gel filtration chromatography.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity±
Column FormatAnalytical Column
Column TypeHydrophobic Interaction
Green FeaturesFSC
Max. Pressure3500 psi (240 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Product TypeAnalytical Column
QuantityEach
Stationary PhaseAmide/Ethyl
Surface Area100 m²/g
Temperature40°C
Diameter (Metric)7.8 mm
Length (Metric)75 mm
MaterialStainless Steel
Particle Size5 μm
PhaseHydrophobic Interaction
Pore Size300 Å
Product LineProPac
TechniqueBio-LC
pH2.5 to 7.5
Unit SizeEach