Dionex™ IonPac™ AS23 IC Columns
Actual product may vary
Dionex™ IonPac™ AS23 IC Columns
Dionex™ IonPac™ AS23 IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS23 IC Columns

Green features
High capacity, anion-exchange column that uses an isocratic carbonate/bicarbonate eluent for analysis of trace bromate and other oxyhalides in diverse environmental samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
064145250 mm2 mmAnalytical Column
06414350 mm2 mmGuard Column
06414750 mm4 mmGuard Column
064149250 mm4 mmAnalytical Column
Catalog number 064145
Price (USD)
1,930.00
Each 1
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each 1

Measure trace bromate and other oxyhalides in drinking water, ground water, wastewater, and other diverse sample matrices with the Thermo Scientific™ Dionex™ IonPac™ AS23 carbonate eluent anion-exchange column. The column is specifically designed and recommended for the determination of trace bromate, a toxic byproduct of ozone disinfection, in drinking water matrices. The column's high capacity and selectivity facilitate the determination of bromate in drinking water at the low-μg/L level in the presence of high concentration of chloride, sulfate and carbonate. The Dionex IonPac AS23 columns meet the performance requirements of U.S. EPA Methods 300.0 and 300.1.

Analysis of Trace Level Bromate in Drinking Water
  • Compliance monitoring of common anions and oxyhalides in drinking water as specified in U.S. EPA Method 300.0 and 300.1
  • Simple analysis of trace bromate in drinking water matrices using an isocratic carbonate/bicarbonate eluent with suppressed conductivity detection
  • Optimized selectivity to ensure low ppb determination of bromate in the presence of very high concentrations of chloride, sulfate, and carbonate
  • Separates oxyhalides and common inorganic anions in drinking water, groundwater, wastewater and other diverse sample matrices
  • High capacity for the analysis of most drinking water without the use of sample pretreatment or preconcentration
  • Solvent compatibility for easy column cleanup after the analysis of samples with hydrophobic components
Multiple Formats Available for Application Flexibility
  • 4 x 250 mm, 2 x 250 mm analytical columns
  • 4 x 50 mm, 2 x 50 mm guard columns
  • Longer 250 mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Note: For simple, accurate eluent preparation, use the Dionex IonPac AS23 Eluent Concentrate.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity80 μeq
Certifications/ComplianceU.S. EPA Methods 300.0 and 300.1
Column Capacity80 μeq
Column ChemistryCarbonate/Bicarbonate
Column ConstructionPEEK
Crosslinking55% DVB
Flow Rate0.25 to 0.5 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)IonPac AG23 Guard Column
Green FeaturesFSC
HydrophobicityUltralow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure3000 psi (207 bar)
Packing MaterialPolymeric
Particle Size6 μm
Pore Size2000 &‌Aring;
QuantityEach 1
Solvent Compatibility0 to 100% HPLC Solvents
Stationary PhaseIonPac AS23
Column TypeAnalytical Column
Diameter (Metric)2 mm
Length (Metric)250 mm
Unit SizeEach 1