Dionex™ IonPac™ AS24 Analytical & Guard Columns
Actual product may vary
Dionex™ IonPac™ AS24 Analytical & Guard Columns
Dionex™ IonPac™ AS24 Analytical & Guard Columns
Dionex™ IonPac™ AS24 Analytical & Guard Columns
Dionex™ IonPac™ AS24 Analytical & Guard Columns
Dionex™ IonPac™ AS24 Analytical & Guard Columns
Dionex™ IonPac™ AS24 Analytical & Guard Columns
Thermo Scientific™

Dionex™ IonPac™ AS24 Analytical & Guard Columns

Green features
Separate haloacetic acids and bromate in drinking water samples prior to detection with mass spectrometry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Column Type
06415150 mmGuard Column
064153250 mmAnalytical Column
Catalog number 064151
Price (USD)
631.00
Each 1
Length (Metric):
50 mm
Column Type:
Guard Column
Price (USD)
631.00
Each 1
Separate haloacetic acids and bromate in drinking water samples with the Thermo Scientific™ Dionex™ IonPac™ AS24 Analytical & Guard Columns. This high-capacity hydroxide-selective anion-exchange column was developed specifically for the separation of haloacetic acids and bromate in drinking water prior to detection with mass spectrometry. The 2 x 250 mm format is the column specified in U.S. EPA Method 557. The capacity and selectivity of the Dionex IonPac AS24 column enables analysis of haloacetic acids in drinking water at low-μg/L (ppb) concentrations. The column is also ideal for the development of specialized applications, providing excellent separation of environmental anions including inorganic anions, oxyhalides, oxyanions, and organic acids in complex matrices.
Superior Chromatographic Performance
  • Specified for U.S. EPA Method 557 (2x250mm)
  • High-capacity 140μeq per column (2 x 250 mm) allows injection of complex matrices without overloading the column while maintaining excellent peak shapes for lower concentration analytes
  • Sample pretreatment is not required when determining haloacetic acids in high-ionic strength matrices
  • Can be used as second dimension column in 2D method for bromate in U.S. EPA Method 302.0 (2 x 250 mm)
  • Ideal for development of specialized applications using Reagent-Free™ Ion Chromatography (RFIC™) systems with gradient elution
  • Can be operated at ambient or elevated temperatures; selectivity is optimized for a 15ºC operating temperature to ensure reproducible recovery for haloacetic acids
  • Compatibility with HPLC organic solvents provides operational flexibility and ease of cleanup
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceU.S. EPA Method 557
Column ChemistryHydroxide
For Use With (Application)Separation of Haloacetic Acids and Bromate in Drinking Water, Anion Exchange
For Use With (Equipment)Separation of Haloacetic Acids and Bromate in Drinking Water
Green FeaturesFSC
Max. Pressure3000 psi (207 bar)
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Column TypeGuard Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)50 mm
Unit SizeEach 1