Dionex&trade; ICS-5000<sup>+</sup> TC Thermal Compartment Stabilizers
Actual product may vary
Thermo Scientific™
Dionex™ ICS-5000+ TC Thermal Compartment Stabilizers
Thermo Scientific™ Dionex™ ICS-5000+ Thermal Stabilizer is installed within the ICS-5000+ Thermal Compartment (TC), which supports up to two stabilizers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
0645484 mm columns, PEEK
0646502 mm columns, PEEK
Catalog number 064548
Price (USD)
-
Description:
4 mm columns, PEEK
Thermo Scientific™ Dionex™ ICS-5000+ Thermal Stabilizer is installed within the ICS-5000+ Thermal Compartment (TC), which supports up to two stabilizers.
Thermal stabilizers ensure that the temperature of the stationary phase remains constant over the total column length, and the column and eluent have the same temperature during the analysis.
  • Available in microbore or standard bore formats
Specifications
Bore SizeStandard; 0.25 mm
Description4 mm columns, PEEK
For Use With (Application)Ion Chromatography
For Use With (Equipment)Dionex ICS-5000+ Thermal Compartment (TC)
TypeTemperature Stabilizer
Unit SizeEach