Dionex™ IonPac™ AS17-C IC Columns
Actual product may vary
Dionex™ IonPac™ AS17-C IC Columns
Dionex™ IonPac™ AS17-C IC Columns
Dionex™ IonPac™ AS17-C IC Columns
Dionex™ IonPac™ AS17-C IC Columns
Dionex™ IonPac™ AS17-C IC Columns
Dionex™ IonPac™ AS17-C IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS17-C IC Columns

Green features
Quickly determine common inorganic anions in high-purity water matrices.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
066296250 mm2 mmAnalytical Column
066294250 mm4 mmAnalytical Column
06629750 mm2 mmGuard Column
06629550 mm4 mmGuard Column
Catalog number 066296
Price (USD)
1,930.00
Each 1
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each 1
Quickly determine common inorganic anions in high-purity water matrices with the Thermo Scientific™ Dionex IonPac AS17-C column. This low-capacity column is optimized for fast gradient separation of inorganic anions in simple sample matrices. It is also ideal for fast gradient separation of sulfur species, including sulfite, sulfate, and thiosulfate. The Dionex IonPac AS17C column is the recommended replacement for all Dionex IonPac AS17 column anion applications.
Make Gradient Separations as Easy as Isocratic Runs

The Dionex IonPac AS17-C column is designed to be used in conjunction with the Dionex Eluent Generator. The Dionex Eluent Generator automatically produces potassium hydroxide eluents from water, making gradient separations as easy as isocratic runs. Using the Dionex Eluent Generator under gradient conditions, common inorganic anions can easily be separated in a variety of sample matrices including high purity water, drinking water, wastewater, process streams, and scrubber solutions.

Achieve Superior Chromatographic Performance
  • Fast gradient separation of inorganic anions in high purity water matrices
  • Easy suppression of hydroxide eluent to a very low background with the Thermo Scientific™ Dionex™ Anion Self-Regenerating Suppressor (ASRS™ 300) for trace level determinations (including fluoride) even with a large loop injection
  • Superior retention and quantification of fluoride, acetate, propionate, and formate
  • Meets performance requirements specified in U.S. EPA Method 300.0 (A)
  • Operates at ambient or elevated temperatures
  • Column selectivity is optimized for operating temperature of 30°C to ensure reproducible results
  • Compatible with Reagent-Free™ Ion Chromatography (RFIC™) systems
  • Compatible with organic solvents to enhance analyte solubility, modify column selectivity, or for effective column clean-up
Note: The Thermo Scientific™ Dionex™ IonPac™ AS18 column is the recommended hydroxide-selective column for determination of common anions in diverse sample matrices.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity7.5 μeq
Certifications/ComplianceU.S. EPA Method 300.0 (A)
Column Capacity7.5 μeq
Column ChemistryHydroxide
Column ConstructionPEEK
Crosslinking55% DVB
Diameter (Metric) Latex75 nm
Flow Rate0.25 to 0.75 mL/min.
For Use With (Application)Anion Exchange
For Use With (Equipment)ICS-2000, ICS-3000 IC System
Green FeaturesFSC
HydrophobicityLow
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Latex Crosslinking6%
Max. Pressure4000 psi (275 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size10.5 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Solvents0 to 100% of HPLC Solvents
Stationary PhaseIonPac AS17-C
Column TypeAnalytical Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)250 mm
pH0 to 14
Unit SizeEach 1