Dionex™ ICS-6000 AutoPrep for AM Automation Manager
Thermo Scientific™
Dionex™ ICS-6000 AutoPrep for AM Automation Manager
Use AutoPrep for trace analysis to facilitate large volume sampling with automated sample preparation in Dionex ICS-6000 HPIC systems with Automation Manager.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
066342AutoPrep Loops Kit
075953AutoPrep Kit with Pre-Plumbed Automation Manager
Catalog number 066342
Price (USD)
943.00
Each 1
Description:
AutoPrep Loops Kit
Price (USD)
943.00
Each 1
Facilitate trace analysis by combining large volume sampling with automated sample preparation with the Thermo Scientific™ Dionex™ ICS-6000 AutoPrep for AM Automation Manager.
Analysis of anions and cations at levels lower than 500 ppt can present unique challenges with respect to sample preparation and contamination. As lower detection limits are required, environmental factors, material compatibility and contamination are increasingly problematic.
  • Automatically calibrates the ICS-5000+/ICS-6000 Reagent-Free™ system
  • Automatically concentrates and analyzes samples
  • Frees the analyst from performing manual sample preparation and standard dilution
  • Improves consistency
  • Minimizes contamination that can interfere with trace analysis
  • Eliminates sample contact with pump components in AS-HV pull mode
AutoPrep System without Automation Manager (Mfr. No. 066342)
  • For ICS-5000+/ICS-6000 with Automation Manager already installed
  • Includes sample and standard loops
AutoPrep System with Automation Manager (Mfr. No. 066343)
  • Includes pre-plumbed Automation Manager, sample and standard loops, 10-port valve
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionAutoPrep Loops Kit
For Use With (Equipment)ICS-5000+/ICS-6000 Ion Chromatography System with automation manager already installed
IncludesSample loops, standard loops
QuantityEach 1
Product TypeAutoPrep Kit
Unit SizeEach 1